missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl 98.0+%, TCI America™

Catalog No. B41661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41661G 1 g
B41665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41661G Supplier TCI America Supplier No. B41661G

Chemical Identifiers

CAS 207611-87-8
Molecular Formula C24H32B2O4
Molecular Weight (g/mol) 406.14
MDL Number MFCD08669545
InChI Key INNVTCAYPFZCAL-UHFFFAOYSA-N
Synonym 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl, 4,4'-biphenyldiboronic acid dipinacol ester, 4,4'-biphenyldiboronic acid pinacol ester, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, biphenyl-4,4'-diboronic acid bis pinacol ester, 4,4'-biphenyldiboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl-4-yl-1,3,2-dioxaborolane, amtb257, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl biphenyl
PubChem CID 10525359
IUPAC Name 4,4,5,5-tetramethyl-2-[4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-yl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 262°C
Color White
Quantity 1 g
Formula Weight 406.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl