missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(2-benzoxazolyl)stilbene 97.0+%, TCI America™

Catalog No. B272925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B272925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B272925G Supplier TCI America Supplier No. B272925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1533-45-5
Molecular Formula C28H18N2O2
Molecular Weight (g/mol) 414.464
InChI Key ORACIQIJMCYPHQ-MDZDMXLPSA-N
Synonym 2,2'-1,2-ethenediyldi-4,1-phenylene bisbenzoxazole, fluorescent brightener ob-1, unii-128by3erd5, 1,2-bis 4-benzo d oxazol-2-yl phenyl ethene, fluorescent brightener 393, 2,2'-vinylenedi-p-phenylene bisbenzoxazole, 4,4'-bis 2-benzoxazolyl stilbene, benzoxazole, 2,2'-1,2-ethenediyldi-4,1-phenylene bis, 2-4-e-2-4-1,3-benzoxazol-2-yl phenyl ethenyl phenyl-1,3-benzoxazole, 2,2'-vinylenedi-4-phenylene bis benzoxazole
PubChem CID 5702717
IUPAC Name 2-[4-[(E)-2-[4-(1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl]-1,3-benzoxazole
SMILES C1=CC=C2C(=C1)N=C(O2)C3=CC=C(C=C3)C=CC4=CC=C(C=C4)C5=NC6=CC=CC=C6O5

Specifications

Melting Point 355°C
Color Yellow
Quantity 25 g
Formula Weight 414.46
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(2-benzoxazolyl)stilbene
EINECSNumber (5)-0168
TSCA Yes