missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bipyridyl 98.0+%, TCI America™

Catalog No. p-7118464
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B046925G 25 g
B0469100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B046925G Supplier TCI America Supplier No. B046925G
Only null left

Chemical Identifiers

CAS 553-26-4
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006416
InChI Key MWVTWFVJZLCBMC-UHFFFAOYSA-N
Synonym 4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridyl, 4-4-pyridyl pyridine, 4,4'-dipyridine, 4,4-bipyridyl, 4,4-dipyridyl, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb
PubChem CID 11107
ChEBI CHEBI:30985
IUPAC Name 4,4'-bipyridine
SMILES C1=CC(=CC=N1)C1=CC=NC=C1

Specifications

Melting Point 112°C
Color White-Yellow
Boiling Point 161°C
Quantity 25 g
UN Number 2811
Formula Weight 156.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bipyridyl
EINECSNumber (5)-3723
RTECSNumber DW1760000
TSCA Yes