missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bipiperidine 98.0+%, TCI America™

Catalog No. p-7123283
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39831G 1 g
B39835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39831G Supplier TCI America Supplier No. B39831G

Chemical Identifiers

CAS 15336-72-8
Molecular Formula C10H20N2
Molecular Weight (g/mol) 168.284
MDL Number MFCD01104315
InChI Key PRNRUOJLUPUJDN-UHFFFAOYSA-N
PubChem CID 736050
IUPAC Name 4-piperidin-4-ylpiperidine
SMILES C1CNCCC1C2CCNCC2

Specifications

Melting Point 171°C
Color White-Yellow
Quantity 1 g
Formula Weight 168.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bipiperidine