missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Biphenylyl)-4-oxobutyric acid, 96%

Catalog No. p-7040711
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6055703 1 g
AAH6055706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6055703 Supplier Thermo Scientific Chemicals Supplier No. H6055703
Only null left

CAS: 36330-85-5 | C16H14O3 | 254.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36330-85-5
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.285
MDL Number MFCD00056701
InChI Key ZPAKPRAICRBAOD-UHFFFAOYSA-N
Synonym fenbufen, lederfen, cinopal, bufemid, napanol, 4-4-biphenylyl-4-oxobutyric acid, 3-4-phenylbenzoyl propionic acid, 3-4-biphenylylcarbonyl propionic acid, 4-biphenyl-4-yl-4-oxobutanoic acid, gamma-oxo 1,1'-biphenyl-4-butanoic acid
PubChem CID 3335
ChEBI CHEBI:31599
IUPAC Name 4-oxo-4-(4-phenylphenyl)butanoic acid
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CCC(=O)O

Specifications

Melting Point 184°C to 187°C
Quantity 1 g
UN Number UN2811
Formula Weight 254.28
Percent Purity 96%
Chemical Name or Material 4-(4-Biphenylyl)-4-oxobutyric acid
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 252-979-0
RTECSNumber DV1761000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only