missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bicyclohexanol (mixture of isomers) 98.0+%, TCI America™

Catalog No. p-7119163
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
B162125G 25 g
B1621500G 500 g
2 options

Catalog No. B162125G

Supplier: TCI America B162125G

Only null left

Chemical Identifiers

CAS 20601-38-1
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00012056
InChI Key MZXNOAWIRQFYDB-UHFFFAOYSA-N
PubChem CID 88608
IUPAC Name 4-(4-hydroxycyclohexyl)cyclohexan-1-ol
SMILES C1CC(CCC1C2CCC(CC2)O)O

Specifications

Color White
Quantity 25 g
Synonym 1,1′C-Bicyclohexyl-4,4′C-diol, 4,4′C-Dihydroxydicyclohexyl
Formula Weight 198.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bicyclohexanol (mixture of isomers)