missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Azodianiline, 95%

Catalog No. AC401580250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC401580250 25 g Glass bottle
AC401580050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC401580250 Supplier Thermo Scientific Chemicals Supplier No. 401580250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 538-41-0 | C12H12N4 | 212.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 538-41-0
Molecular Formula C12H12N4
Molecular Weight (g/mol) 212.26
MDL Number MFCD00041892
InChI Key KQIKKETXZQDHGE-UHFFFAOYSA-N
Synonym 4,4'-azodianiline, p-azoaniline, p-diaminoazobenzene, 4,4'-diaminoazobenzene, azodianiline, p'-amino-p-aminoazobenzene, 4,4'-azobisbenzenamine, benzenamine, 4,4'-azobis, 4,4-azodianiline, 4,4'-diazenediylbisaniline
PubChem CID 10855
IUPAC Name 4-[(4-aminophenyl)diazenyl]aniline
SMILES NC1=CC=C(C=C1)N=NC1=CC=C(N)C=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 245°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 2979
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,very soluble in benzene and chloroform
Formula Weight 212.25
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4, 4'-Azodianiline, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
IF exposed or concerned: Get medical advice/attention.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,ha
EINECSNumber 208-690-7
RTECSNumber BW7450000
TSCA TSCA

RUO – Research Use Only