missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 4,4'-Azobis(4-cyanovaleric acid)

Catalog No. 11590100G
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11590100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11590100G Supplier Sigma Aldrich Supplier No. 11590100G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,4'-Azobis(4-cyanovaleric acid)

Specifications

Melting Point 118°C to 125°C (dec.) (lit.)
CAS 2638-94-0
Molecular Formula C12H16N4O4
Linear Formula HOCOCH2CH2C(CH3)(CN)N=NC(CH3)(CN)CH2CH2COOH
MDL Number MFCD00002799
Synonym 4,4 -Azobis(4-cyanopentanoic acid); ABCVA; ACVA
Quantity 100 g
Molecular Weight (g/mol) 280.28
Percent Purity ≥98.0% (T)