missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 4-(4-Anilinophenylazo)benzenesulfonic acid sodium salt, indicator grade
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Color | Ochre to Orange |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Quantity | 25 g |
| Merck Index | 15, 9955 |
| Formula Weight | 375.38 |
| Grade | Extra Pure |
| Physical Form | Powder |
| Chemical Name or Material | 4-(4-Anilinophenylazo)benzenesulfonic acid sodium salt |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement: Harmful in contact with skin. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 209-071-4 |
| TSCA | TSCA |
RUO – Research Use Only