missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-(4-Anilinophenylazo)benzenesulfonic acid sodium salt, indicator grade

Catalog No. AC401260250
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC401260250 25 g
AC401261000 100 g
AC401260010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC401260250 Supplier Thermo Scientific Chemicals Supplier No. 401260250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 554-73-4 | C18H14N3NaO3S | 375.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 554-73-4
Molecular Formula C18H14N3NaO3S
Molecular Weight (g/mol) 375.38
MDL Number MFCD00038130
InChI Key MLVYOYVMOZFHIU-UHFFFAOYSA-M
Synonym orange iv, tropaeolin oo, orangeiv, acid orange 5, diphenylamine orange, tropaeolin, unii-vr8q3r288w, tropeolin oo, orange n, solar orange iv
PubChem CID 23668839
IUPAC Name sodium;4-[(4-anilinophenyl)diazenyl]benzenesulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Color Ochre to Orange
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 9955
Formula Weight 375.38
Grade Extra Pure
Physical Form Powder
Chemical Name or Material 4-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-071-4
TSCA TSCA

RUO – Research Use Only