missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Aminophenyl)morpholin-3-one 98.0+%, TCI America™

Catalog No. p-7117992
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26141G 1 g
A26145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26141G Supplier TCI America Supplier No. A26141G

Chemical Identifiers

CAS 438056-69-0
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.22
MDL Number MFCD08236742
InChI Key MHCRLDZZHOVFEE-UHFFFAOYSA-N
Synonym 4-(3-Oxomorpholino)aniline
PubChem CID 11194854
IUPAC Name 4-(4-aminophenyl)morpholin-3-one
SMILES NC1=CC=C(C=C1)N1CCOCC1=O

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 1 g
Formula Weight 192.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Aminophenyl)morpholin-3-one