missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Aminophenoxy)-N-methyl-2-pyridinecarboxamide 98.0+%, TCI America™
$52.19 - $160.30
Chemical Identifiers
| CAS | 284462-37-9 |
|---|---|
| Molecular Formula | C13H13N3O2 |
| Molecular Weight (g/mol) | 243.27 |
| MDL Number | MFCD08692047 |
| InChI Key | RXZZBPYPZLAEFC-UHFFFAOYSA-N |
| Synonym | 4-(4-Aminophenoxy)-N-methylpicolinamide |
| PubChem CID | 16655129 |
| IUPAC Name | 4-(4-aminophenoxy)-N-methylpyridine-2-carboxamide |
| SMILES | CNC(=O)C1=NC=CC(OC2=CC=C(N)C=C2)=C1 |
Chemical Identifiers
| 284462-37-9 | |
| 243.27 | |
| RXZZBPYPZLAEFC-UHFFFAOYSA-N | |
| 16655129 | |
| CNC(=O)C1=NC=CC(OC2=CC=C(N)C=C2)=C1 |
| C13H13N3O2 | |
| MFCD08692047 | |
| 4-(4-Aminophenoxy)-N-methylpicolinamide | |
| 4-(4-aminophenoxy)-N-methylpyridine-2-carboxamide |
Specifications
| 284462-37-9 | |
| C13H13N3O2 | |
| 1 g | |
| RXZZBPYPZLAEFC-UHFFFAOYSA-N | |
| 4-(4-aminophenoxy)-N-methylpyridine-2-carboxamide | |
| 16655129 | |
| ≥98.0% (HPLC,T) | |
| 4-(4-Aminophenoxy)-N-methyl-2-pyridinecarboxamide |
| 118°C | |
| MFCD08692047 | |
| 4-(4-Aminophenoxy)-N-methylpicolinamide | |
| CNC(=O)C1=NC=CC(OC2=CC=C(N)C=C2)=C1 | |
| 243.27 | |
| 243.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : No