missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Acetyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog No. AAH5977103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5977103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5977103 Supplier Thermo Scientific Chemicals Supplier No. H5977103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1218791-38-8
Molecular Formula C18H27BN2O3
Molecular Weight (g/mol) 330.235
MDL Number MFCD13195756
InChI Key VRZVSHHLWMAHAZ-UHFFFAOYSA-N
Synonym 4-4-acetyl-1-piperazinyl phenylboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazin-1-yl ethanone, 4-4-acetylpiperazino phenylboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1-piperazinyl ethanone, 4-4-acetyl-1-piperazinyl phenylboronicacidpinacolester, 4-4-acetylpiperazino phenylboronic acid, pinacol ester, 4-4-acetyl-1-piperazinyl benzeneboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazin-1-yl ethan-1-one
PubChem CID 53217141
IUPAC Name 1-[4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazin-1-yl]ethanone
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCN(CC3)C(=O)C

Specifications

Quantity 1 g
Formula Weight 330.23
Percent Purity 95%
Chemical Name or Material 4-(4-Acetyl-1-piperazinyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only