missing translation for 'onlineSavingsMsg'
Learn More

4-(4,6-Dichloro-2-pyrimidyl)morpholine 98.0+%, TCI America™

Catalog No. p-7128086
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40691G 1 g
D40695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40691G Supplier TCI America Supplier No. D40691G
Only null left

Chemical Identifiers

CAS 10397-13-4
Molecular Formula C8H9Cl2N3O
Molecular Weight (g/mol) 234.08
MDL Number MFCD05022359
InChI Key OXCOCPRVQUEIOL-UHFFFAOYSA-N
PubChem CID 2772068
IUPAC Name 4-(4,6-dichloropyrimidin-2-yl)morpholine
SMILES ClC1=CC(Cl)=NC(=N1)N1CCOCC1

Specifications

Melting Point 141°C
Color White
Quantity 1 g
Synonym 4-4,6-dichloropyrimidin-2-yl morpholine, 4-4,6-dichloro-pyrimidin-2-yl-morpholine, 4-4,6-dichloro-2-pyrimidyl morpholine, 2-morpholino-4,6-dichloropyrimidine, 4,6-dichloro-2-morpholinopyrimidine, 2-4-morpholino-4,6-dichloropyrimidine, morpholine,4-4,6-dichloro-2-pyrimidinyl, morpholine, 4-4,6-dichloro-2-pyrimidinyl, 4-4,6-dichloro-2-pyrimidinyl morpholine, pubchem21502
Formula Weight 234.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,6-Dichloro-2-pyrimidyl)morpholine