missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(tetrahydrofuran-3-yl)-1,3,2-dioxaborolane, 97%

Catalog No. AC431100010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431100010 1 g Glass bottle
AC431100050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC431100010 Supplier Thermo Scientific Chemicals Supplier No. 431100010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 331958-90-8 | C10H19BO3 | 198.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 331958-90-8
Molecular Formula C10H19BO3
Molecular Weight (g/mol) 198.07
MDL Number MFCD09878900
InChI Key KEBDNHHRQNZXMA-UHFFFAOYNA-N
Synonym 4,4,5,5-tetramethyl-2-tetrahydrofuran-3-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-oxolan-3-yl-1,3,2-dioxaborolane, tetrahydrofuran-3-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-tetrahydro-3-furanyl-1,3,2-dioxaborolane, tetrahydrofuran-3-ylboronic acid pinacol ester, tetrahydrofuran-3-yl boronic acid pinacol ester, 2-tetrahydrofuran-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl tetrahydrofuran
PubChem CID 11148297
IUPAC Name 4,4,5,5-tetramethyl-2-(oxolan-3-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1CCOC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 198.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(tetrahydrofuran-3-yl)-1,3,2-dioxaborolane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

RUO – Research Use Only