missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(p-tolyl)-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. T28881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28881G 1 g
T28885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28881G Supplier TCI America Supplier No. T28881G
Only null left

Chemical Identifiers

CAS 195062-57-8
Molecular Formula C13H19BO2
Molecular Weight (g/mol) 218.103
MDL Number MFCD03453662
InChI Key GKSSEDDAXXEPCP-UHFFFAOYSA-N
Synonym p-Tolylboronic Acid Pinacol Ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)toluene
PubChem CID 2774016
IUPAC Name 4,4,5,5-tetramethyl-2-(4-methylphenyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C

Specifications

Melting Point 54°C
Color White
Quantity 1 g
Formula Weight 218.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(p-tolyl)-1,3,2-dioxaborolane