missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(o-tolyl)-1,3,2-dioxaborolane 97.0+%, TCI America™

Catalog No. T31951G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31951G 1 g
T31955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31951G Supplier TCI America Supplier No. T31951G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 195062-59-0
Molecular Formula C13H19BO2
Molecular Weight (g/mol) 218.103
MDL Number MFCD06795658
InChI Key NUFSJKMRPYGNHV-UHFFFAOYSA-N
Synonym o-Tolylboronic Acid Pinacol Ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)toluene
PubChem CID 10704008
IUPAC Name 4,4,5,5-tetramethyl-2-(2-methylphenyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C

Specifications

Color Yellow
Quantity 1 g
Formula Weight 218.10
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(o-tolyl)-1,3,2-dioxaborolane