missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(5-methylfuran-2-yl)-1,3,2-dioxaborolane 97.0+%, TCI America™

Catalog No. p-7137803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35821G 1 g
T35825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35821G Supplier TCI America Supplier No. T35821G

Chemical Identifiers

CAS 338998-93-9
Molecular Formula C11H17BO3
Molecular Weight (g/mol) 208.06
MDL Number MFCD03094689
InChI Key FNPZFZKLYGWKLH-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-5-methylfuran-2-yl-1,3,2-dioxaborolane, 5-methylfuran-2-boronic acid pinacol ester, 2-methylfurane-5-boronic acid pinacol ester, 2-methylfuran-5-boronic acid pinacol ester, 2-methyl-furyl-5-pinacolato boron, 5-methyl-2-furanboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-5-methyl-2-furyl-1,3,2-dioxaborolane, 5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxoborolan-2-yl furan, 4,4,5,5-tetramethyl-2-5-methyl-2-furanyl-1,3,2-dioxaborolane, 5-methylfuran-2-yl boronic acid pinacol ester
PubChem CID 2736881
IUPAC Name 4,4,5,5-tetramethyl-2-(5-methylfuran-2-yl)-1,3,2-dioxaborolane
SMILES CC1=CC=C(O1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 1 g
Formula Weight 208.06
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(5-methylfuran-2-yl)-1,3,2-dioxaborolane
TSCA No
Recommended Storage Refrigerator