missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-[4-(thien-2-ylmethyl)phenyl]-1,3,2-dioxaborolane, 97%, Thermo Scientific™

Catalog No. p-3882134
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC70639CB 250mg Amber Glass Bottle
CC70639DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC70639CB Supplier Thermo Scientific Chemicals Supplier No. CC70639CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1007847-76-8
Molecular Formula C17H21BO2S
Molecular Weight (g/mol) 300.223
MDL Number MFCD11841077
InChI Key OPJUSDABOPHYPQ-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-4-thien-2-ylmethyl phenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-4-thiophen-2-ylmethyl phenyl-1,3,2-dioxaborolane, 4-thien-2-ylmethyl benzeneboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-4-thiophen-2-yl methyl phenyl-1,3,2-dioxaborolane
PubChem CID 43811052
IUPAC Name 4,4,5,5-tetramethyl-2-[4-(thiophen-2-ylmethyl)phenyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC3=CC=CS3

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 300.23
Percent Purity 97%
Chemical Name or Material 4,4,5,5-Tetramethyl-2-[4-(thien-2-ylmethyl)phenyl]-1,3,2-dioxaborolane
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.