missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-[4-(thien-2-ylmethyl)phenyl]-1,3,2-dioxaborolane, 97%, Thermo Scientific™

Numéro de catalogue. CC70639CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC70639CB 250mg Amber Glass Bottle
CC70639DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC70639CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC70639CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 1007847-76-8
Formule moléculaire C17H21BO2S
Poids moléculaire (g/mol) 300.223
Numéro MDL MFCD11841077
Clé InChI OPJUSDABOPHYPQ-UHFFFAOYSA-N
Synonyme 4,4,5,5-tetramethyl-2-4-thien-2-ylmethyl phenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-4-thiophen-2-ylmethyl phenyl-1,3,2-dioxaborolane, 4-thien-2-ylmethyl benzeneboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-4-thiophen-2-yl methyl phenyl-1,3,2-dioxaborolane
PubChem CID 43811052
Nom de l’IUPAC 4,4,5,5-tetramethyl-2-[4-(thiophen-2-ylmethyl)phenyl]-1,3,2-dioxaborolane
SOURIRES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC3=CC=CS3

Spécifications

Nom ou substance chimique 4,4,5,5-Tetramethyl-2-[4-(thien-2-ylmethyl)phenyl]-1,3,2-dioxaborolane
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 300.23
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.