missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(2-naphthyl)-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. p-7137682
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32981G 1 g
T32985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32981G Supplier TCI America Supplier No. T32981G

Chemical Identifiers

CAS 256652-04-7
Molecular Formula C16H19BO2
Molecular Weight (g/mol) 254.136
MDL Number MFCD05663868
InChI Key SPPZBAGKKBHZRW-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane, naphthalene-2-boronic acid pinacol ester, naphthalene-2-boronic acid, pinacol ester, 2-naphthaleneboronic acid, pinacol ester, 2-naphth-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthalenyl, 2-naphthaleneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-naphthalenyl-1,3,2-dioxaborolane, amtb668
PubChem CID 10753409
IUPAC Name 4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC=CC=C3C=C2

Specifications

Melting Point 66°C
Color White
Quantity 1 g
Formula Weight 254.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(2-naphthyl)-1,3,2-dioxaborolane