missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(2-methyl-3-furyl)-1,3,2-dioxaborolane, 90%

Catalog No. AC451520050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451520050 5 g
AC451520010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC451520050 Supplier Thermo Scientific Chemicals Supplier No. 451520050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 864776-02-3 | C11H17BO3 | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 864776-02-3
Molecular Formula C11H17BO3
Molecular Weight (g/mol) 208.06
MDL Number MFCD09879905
InChI Key JMLBPHNESOKSEV-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-2-methylfuran-3-yl-1,3,2-dioxaborolane, 2-methylfuran-3-boronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-2-methyl-3-furanyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane,4,4,5,5-tetramethyl-2-2-methyl-3-furanyl, 4,4,5,5-tetramethyl-2-2-methyl-3-furyl-1,3,2-dioxaborolane, pubchem18438, 2-2-methylfur-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan, 4,4,5,5-tetramethyl-2-2-methyl 3-furyl-1,3,2-dioxaborolane, 3-hydroxy-2,3-dimethylbutan-2-yl hydrogen 2-methylfuran-3-yl boronate
PubChem CID 24229547
IUPAC Name 4,4,5,5-tetramethyl-2-(2-methylfuran-3-yl)-1,3,2-dioxaborolane
SMILES CC1=C(C=CO1)B1OC(C)(C)C(C)(C)O1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 89% min. (GC)
Quantity 5 g
Formula Weight 208.06
Percent Purity 90%
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(2-methyl-3-furyl)-1,3,2-dioxaborolane
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only