missing translation for 'onlineSavingsMsg'
Learn More

4,4',4″-Tris(carbazol-9-yl)triphenylamine, 99%

Catalog No. p-7077778
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459180010 1 g
AC459180050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459180010 Supplier Thermo Scientific Chemicals Supplier No. 459180010

CAS: 139092-78-7 | C54H36N4 | 740.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139092-78-7
Molecular Formula C54H36N4
Molecular Weight (g/mol) 740.91
MDL Number MFCD03093250
InChI Key AWXGSYPUMWKTBR-UHFFFAOYSA-N
Synonym tris 4-9h-carbazol-9-yl phenyl amine, tcta, tcata, 4,4',4-tris carbazol-9-yl-triphenylamine, 4,4',4-tri-9-carbazolyltriphenylamine, tris 4-carbazoyl-9-ylphenyl amine, tris 4-carbazol-9-ylphenyl amine, 4-carbazol-9-yl-n,n-bis 4-carbazol-9-yl phenyl aniline, 4,4',4-tris carbaz, 4-carbazol-9-yl-n,n-bis 4-carbazol-9-ylphenyl aniline
PubChem CID 9962045
IUPAC Name 4-carbazol-9-yl-N,N-bis(4-carbazol-9-ylphenyl)aniline
SMILES C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=C(C=C1)N(C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 297.0°C to 300.0°C
Elemental Analysis 86.23 to 88.85% Carbon 7.45 to 7.68% Nitrogen
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 740.92
Percent Purity 99%
Chemical Name or Material 4,4',4''-Tris(carbazol-9-yl)triphenylamine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw

RUO – Research Use Only