missing translation for 'onlineSavingsMsg'
Learn More

4,4',4″-Tris[2-naphthyl(phenyl)amino]triphenylamine 98.0+%, TCI America™

Catalog No. t32331g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T32331G Supplier TCI America Supplier No. T32331G

Chemical Identifiers

CAS 185690-41-9
Molecular Formula C66H48N4
Molecular Weight (g/mol) 897.138
MDL Number MFCD01310662
InChI Key KDOQMLIRFUVJNT-UHFFFAOYSA-N
Synonym n1-naphthalen-2-yl-n4,n4-bis 4-naphthalen-2-yl phenyl amino phenyl-n1-phenylbenzene-1,4-diamine, 4,4',4-tris 2-naphthyl phenyl amino triphenylamine, tris 4-2-naphthylphenylamino phenyl amine, 4,4',4-tris n-naphth-2-yl-n-phenylamino triphenylamine, 4,4',4-tris n-2-naphthyl-n-phenyl-amino-triphenylamine, n1,n1-bis 4-naphthalen-2-yl phenyl amino phenyl-n4-naphthalen-2-yl-n4-phenylbenzene-1,4-diamine, 2t-nata, 4,4',4-tris 2-naph, 4,4,4-tris 2-naphthyl phenyl amino triphenylamine, 4,4',4'-tris 2-naphthyl phenyl amino triphenylamine
PubChem CID 16184079
IUPAC Name 4-N-naphthalen-2-yl-1-N,1-N-bis[4-(N-naphthalen-2-ylanilino)phenyl]-4-N-phenylbenzene-1,4-diamine
SMILES C1=CC=C(C=C1)N(C2=CC=C(C=C2)N(C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC6=CC=CC=C6C=C5)C7=CC=C(C=C7)N(C8=CC=CC=C8)C9=CC1=CC=CC=C1C=C9)C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 247°C
Color Yellow
Quantity 1 g
Formula Weight 897.14
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′,4′′-Tris[2-naphthyl(phenyl)amino]triphenylamine