missing translation for 'onlineSavingsMsg'
Learn More

4,4',4″-Trimethyltriphenylamine, 98%

Catalog No. p-7055552
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6416203 1 g
AAH6416206 5 g
AAH6416214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6416203 Supplier Thermo Scientific Chemicals Supplier No. H6416203
Only null left

CAS: 1159-53-1 | C21H21N | 287.406 g/mol

Used for proteomics research. Also employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for proteomics research. Also employed as a intermediate for pharmaceutical.

Solubility
Soluble in chloroform and tetrahydrofuran (THF).

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 1159-53-1
Molecular Formula C21H21N
Molecular Weight (g/mol) 287.406
MDL Number MFCD00674043
InChI Key YXYUIABODWXVIK-UHFFFAOYSA-N
Synonym tri-p-tolylamine, 4,4',4-trimethyltriphenylamine, 4,4,4-trimethyltriphenylamine, tri p-tolyl amine, benzenamine, 4-methyl-n,n-bis 4-methylphenyl, 4-methyl-n,n-bis 4-methylphenyl aniline, tris 4-methylphenyl amine, n,n-di-p-tolyl-p-toluidine, n,n,n-tri p-tolyl amine, trip-tolylamine
PubChem CID 70873
IUPAC Name 4-methyl-N,N-bis(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C

Specifications

Melting Point 114°C to 118°C (literature)
Color White
Quantity 1 g
Beilstein 2215309
Solubility Information Soluble in chloroform and tetrahydrofuran (THF).
Formula Weight 287.41
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4,4',4''-Trimethyltriphenylamine
EINECSNumber 214-595-1
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only