missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluorobutyric Acid 98.0+%, TCI America™

Catalog No. T26705G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26705G 5 g
T26701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26705G Supplier TCI America Supplier No. T26705G

Chemical Identifiers

CAS 406-93-9
Molecular Formula C4H5F3O2
Molecular Weight (g/mol) 142.077
MDL Number MFCD00077604
InChI Key WTUCTMYLCMVYEX-UHFFFAOYSA-N
Synonym 4,4,4-trifluorobutyric acid, 4,4,4-trifluoro-butyric acid, butanoic acid, 4,4,4-trifluoro, zlchem 266, acmc-1ai6e, 4,4,4-trifluorobutyricacid, trifluoropropylcarboxylic acid, 4,4,4 trifluorobutyric acid, ksc237i6h, 3-trifluoromethylpropionic acid
PubChem CID 2777085
IUPAC Name 4,4,4-trifluorobutanoic acid
SMILES C(CC(F)(F)F)C(=O)O

Specifications

Melting Point 31°C
Color White
Boiling Point 167°C
Quantity 5 g
UN Number 3261
Formula Weight 142.08
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 4,4,4-Trifluorobutyric Acid