missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-2-methylbutyric acid, 95%, Thermo Scientific™

Catalog No. p-7046921
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1681306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1681306 Supplier Thermo Scientific Chemicals Supplier No. L1681306

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4,4,4-Trifluoro-2-methylbutyric acid, is used as pharma intermediates

Solubility
Soluble in water and ethanol, and insoluble in ether

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 99783-23-0
Molecular Formula C5H7F3O2
Molecular Weight (g/mol) 156.104
MDL Number MFCD00190640
InChI Key OBUKEMPWKGDRJV-UHFFFAOYSA-N
Synonym 2-methyl-4,4,4-trifluorobutyric acid, 4,4,4-trifluoro-2-methyl-butanoic acid, 2-methyl-4,4,4-trifluorobutanoic acid, 4,4,4-trifluoro-2-methylbutyric acid, 2-methyl-4,4,4-trifluorobutanoic acid, butanoic acid, 4,4,4-trifluoro-2-methyl, 4,4,4-tris fluoranyl-2-methyl-butanoic acid
PubChem CID 2775536
IUPAC Name 4,4,4-trifluoro-2-methylbutanoic acid
SMILES CC(CC(F)(F)F)C(=O)O

Specifications

Boiling Point 172°C to 175°C
Refractive Index 1.358
Quantity 5 g
UN Number UN3265
Beilstein 6917955
Solubility Information Soluble in water and ethanol,and insoluble in ether
Formula Weight 156.11
Percent Purity 95%
Chemical Name or Material 4,4,4-Trifluoro-2-methylbutyric acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures