missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-1-phenyl-1,3-butanedione 98.0+%, TCI America™

Catalog No. T043725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T043725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T043725G Supplier TCI America Supplier No. T043725G

Chemical Identifiers

CAS 326-06-7
Molecular Formula C10H7F3O2
Molecular Weight (g/mol) 216.16
MDL Number MFCD00000425
InChI Key VVXLFFIFNVKFBD-UHFFFAOYSA-N
Synonym benzoyl-1,1,1-trifluoroacetone, benzoyltrifluoroacetone, 4,4,4-trifluoro-1-phenyl-1,3-butanedione, benzoyl trifluoroacetyl methane, 3-benzoyl-1,1,1-trifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-phenyl, 4,4,4-trifluoro-1-phenyl-butane-1,3-dione, 1-benzoyl-3,3,3-trifluoroacetone, .omega.-trifluoroacetyl acetophenone, 1-benzoyl-3,3,3-trifluoro-2-propanone
PubChem CID 67589
IUPAC Name 4,4,4-trifluoro-1-phenylbutane-1,3-dione
SMILES FC(F)(F)C(=O)CC(=O)C1=CC=CC=C1

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 224°C
Quantity 25 g
Formula Weight 216.16
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 4,4,4-Trifluoro-1-phenyl-1,3-butanedione