missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,4-Trifluoro-1-(p-tolyl)-1,3-butanedione 98.0+%, TCI America™
$85.75 - $223.68
Chemical Identifiers
| CAS | 720-94-5 |
|---|---|
| Molecular Formula | C11H9F3O2 |
| Molecular Weight (g/mol) | 230.186 |
| MDL Number | MFCD00517909 |
| InChI Key | WRZMHTIRFOFFPY-UHFFFAOYSA-N |
| Synonym | 4,4,4-trifluoro-1-p-tolyl butane-1,3-dione, 4,4,4-trifluoro-1-4-methylphenyl butane-1,3-dione, 1-4-methylphenyl-4,4,4-trifluorobutane-1,3-dione, l-4-methylphenyl-4,4,4-trifluorobutane-1,3-dione, 4,4,4-trifluoro-1-4-methyl phenyl-butane-1,3-dione, 3-4-methylbenzoyl-1,1,1-trifluoroacetone, 4,4,4-trifluoro-1-p-tolylbutane-1,3-dione, 4,4,4-trifluoro-1-p-tolyl-butane-1,3-dione, 1-4-methylphenyl-4,4,4-triflurobutane-1,3-dione, 1,3-butanedione, 4,4,4-trifluoro-1-4-methylphenyl |
| PubChem CID | 550193 |
| IUPAC Name | 4,4,4-trifluoro-1-(4-methylphenyl)butane-1,3-dione |
| SMILES | CC1=CC=C(C=C1)C(=O)CC(=O)C(F)(F)F |
Chemical Identifiers
| 720-94-5 | |
| 230.186 | |
| WRZMHTIRFOFFPY-UHFFFAOYSA-N | |
| 550193 | |
| CC1=CC=C(C=C1)C(=O)CC(=O)C(F)(F)F |
| C11H9F3O2 | |
| MFCD00517909 | |
| 4,4,4-trifluoro-1-p-tolyl butane-1,3-dione, 4,4,4-trifluoro-1-4-methylphenyl butane-1,3-dione, 1-4-methylphenyl-4,4,4-trifluorobutane-1,3-dione, l-4-methylphenyl-4,4,4-trifluorobutane-1,3-dione, 4,4,4-trifluoro-1-4-methyl phenyl-butane-1,3-dione, 3-4-methylbenzoyl-1,1,1-trifluoroacetone, 4,4,4-trifluoro-1-p-tolylbutane-1,3-dione, 4,4,4-trifluoro-1-p-tolyl-butane-1,3-dione, 1-4-methylphenyl-4,4,4-triflurobutane-1,3-dione, 1,3-butanedione, 4,4,4-trifluoro-1-4-methylphenyl | |
| 4,4,4-trifluoro-1-(4-methylphenyl)butane-1,3-dione |
Specifications
| 720-94-5 | |
| C11H9F3O2 | |
| 5 g | |
| WRZMHTIRFOFFPY-UHFFFAOYSA-N | |
| 4,4,4-trifluoro-1-(4-methylphenyl)butane-1,3-dione | |
| 550193 | |
| ≥98.0% (GC) | |
| 4,4,4-Trifluoro-1-(p-tolyl)-1,3-butanedione |
| 47°C | |
| MFCD00517909 | |
| 4,4,4-trifluoro-1-p-tolyl butane-1,3-dione, 4,4,4-trifluoro-1-4-methylphenyl butane-1,3-dione, 1-4-methylphenyl-4,4,4-trifluorobutane-1,3-dione, l-4-methylphenyl-4,4,4-trifluorobutane-1,3-dione, 4,4,4-trifluoro-1-4-methyl phenyl-butane-1,3-dione, 3-4-methylbenzoyl-1,1,1-trifluoroacetone, 4,4,4-trifluoro-1-p-tolylbutane-1,3-dione, 4,4,4-trifluoro-1-p-tolyl-butane-1,3-dione, 1-4-methylphenyl-4,4,4-triflurobutane-1,3-dione, 1,3-butanedione, 4,4,4-trifluoro-1-4-methylphenyl | |
| CC1=CC=C(C=C1)C(=O)CC(=O)C(F)(F)F | |
| 230.186 | |
| 230.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No