missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,4-Trifluoro-1-(2-thienyl)-1,3-butanedione 98.0+%, TCI America™
$42.87 - $741.89
Chemical Identifiers
| CAS | 326-91-0 |
|---|---|
| Molecular Formula | C8H5F3O2S |
| Molecular Weight (g/mol) | 222.18 |
| MDL Number | MFCD00005445 |
| InChI Key | TXBBUSUXYMIVOS-UHFFFAOYSA-N |
| Synonym | 2-thenoyltrifluoroacetone, thenoyltrifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl, 4,4,4-trifluoro-1-thiophen-2-yl butane-1,3-dione, 4,4,4-trifluoro-1-2-thienyl-1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl butane-1,3-dione, perfluoroacetyl 2-thenoyl methane, alpha-thenoyltrifluoroacetone, 1-thenoyl-3,3,3-trifluoroacetone, .alpha.-thenoyltrifluoroacetone |
| PubChem CID | 5601 |
| IUPAC Name | 4,4,4-trifluoro-1-(thiophen-2-yl)butane-1,3-dione |
| SMILES | FC(F)(F)C(=O)CC(=O)C1=CC=CS1 |
Chemical Identifiers
| 326-91-0 | |
| 222.18 | |
| TXBBUSUXYMIVOS-UHFFFAOYSA-N | |
| 5601 | |
| FC(F)(F)C(=O)CC(=O)C1=CC=CS1 |
| C8H5F3O2S | |
| MFCD00005445 | |
| 2-thenoyltrifluoroacetone, thenoyltrifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl, 4,4,4-trifluoro-1-thiophen-2-yl butane-1,3-dione, 4,4,4-trifluoro-1-2-thienyl-1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl butane-1,3-dione, perfluoroacetyl 2-thenoyl methane, alpha-thenoyltrifluoroacetone, 1-thenoyl-3,3,3-trifluoroacetone, .alpha.-thenoyltrifluoroacetone | |
| 4,4,4-trifluoro-1-(thiophen-2-yl)butane-1,3-dione |
Specifications
| 326-91-0 | |
| White-Yellow | |
| C8H5F3O2S | |
| 25 g | |
| TXBBUSUXYMIVOS-UHFFFAOYSA-N | |
| 4,4,4-trifluoro-1-(thiophen-2-yl)butane-1,3-dione | |
| 5601 | |
| ≥98.0% (GC) | |
| 4,4,4-Trifluoro-1-(2-thienyl)-1,3-butanedione |
| 45°C | |
| 104°C | |
| MFCD00005445 | |
| 2-thenoyltrifluoroacetone, thenoyltrifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl, 4,4,4-trifluoro-1-thiophen-2-yl butane-1,3-dione, 4,4,4-trifluoro-1-2-thienyl-1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl butane-1,3-dione, perfluoroacetyl 2-thenoyl methane, alpha-thenoyltrifluoroacetone, 1-thenoyl-3,3,3-trifluoroacetone, .alpha.-thenoyltrifluoroacetone | |
| FC(F)(F)C(=O)CC(=O)C1=CC=CS1 | |
| 222.18 | |
| 222.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes