missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-1-(2-thienyl)-1,3-butanedione 98.0+%, TCI America™

Catalog No. T043825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T043825G 25 g
T0438500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T043825G Supplier TCI America Supplier No. T043825G

Chemical Identifiers

CAS 326-91-0
Molecular Formula C8H5F3O2S
Molecular Weight (g/mol) 222.18
MDL Number MFCD00005445
InChI Key TXBBUSUXYMIVOS-UHFFFAOYSA-N
Synonym 2-thenoyltrifluoroacetone, thenoyltrifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl, 4,4,4-trifluoro-1-thiophen-2-yl butane-1,3-dione, 4,4,4-trifluoro-1-2-thienyl-1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl butane-1,3-dione, perfluoroacetyl 2-thenoyl methane, alpha-thenoyltrifluoroacetone, 1-thenoyl-3,3,3-trifluoroacetone, .alpha.-thenoyltrifluoroacetone
PubChem CID 5601
IUPAC Name 4,4,4-trifluoro-1-(thiophen-2-yl)butane-1,3-dione
SMILES FC(F)(F)C(=O)CC(=O)C1=CC=CS1

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 104°C
Quantity 25 g
Formula Weight 222.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,4-Trifluoro-1-(2-thienyl)-1,3-butanedione