missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione, 98%

Catalog No. AAA1069314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1069314 25 g
AAA1069306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1069314 Supplier Thermo Scientific Chemicals Supplier No. A1069314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 326-90-9 | C8H5F3O3 | 206.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 326-90-9
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD00020935
InChI Key OWLPCALGCHDBCN-UHFFFAOYSA-N
Synonym 4,4,4-trifluoro-1-furan-2-yl butane-1,3-dione, 2-furoyltrifluoroacetone, furoyltrifluoroacetone, 4,4,4-trifluoro-1-2-furyl-1,3-butanedione, perfluoroacetyl 2-furoyl methane, furonyltrifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-furanyl, 4,4,4-trifluoro-1-2-furyl butane-1,3-dione, 2-furoyl trifluoro acetone, 4,4,4-trifluoro-1-2-furanyl-1,3-butanedione
PubChem CID 67594
IUPAC Name 4,4,4-trifluoro-1-(furan-2-yl)butane-1,3-dione
SMILES FC(F)(F)C(=O)CC(=O)C1=CC=CO1

Specifications

Melting Point 20°C to 21°C
Density 1.391
Boiling Point 203°C
Flash Point 87°C (188°F)
Refractive Index 1.528
Quantity 25 g
Beilstein 168085
Formula Weight 206.12
Percent Purity 98%
Chemical Name or Material 4,4,4-Trifluoro-1- (2-furyl)-1,3-butanedione
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 206-315-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only