missing translation for 'onlineSavingsMsg'
Learn More

4,4',4″-Tri-9-carbazolyltriphenylamine 98.0+%, TCI America™

Catalog No. p-7137318
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
T26161G 1 g
T26165G 5 g
2 options

Catalog No. T26161G

Supplier: TCI America T26161G

Only null left

Chemical Identifiers

CAS 139092-78-7
Molecular Formula C54H36N4
Molecular Weight (g/mol) 740.91
MDL Number MFCD03093250
InChI Key AWXGSYPUMWKTBR-UHFFFAOYSA-N
Synonym tris 4-9h-carbazol-9-yl phenyl amine, tcta, tcata, 4,4',4-tris carbazol-9-yl-triphenylamine, 4,4',4-tri-9-carbazolyltriphenylamine, tris 4-carbazoyl-9-ylphenyl amine, tris 4-carbazol-9-ylphenyl amine, 4-carbazol-9-yl-n,n-bis 4-carbazol-9-yl phenyl aniline, 4,4',4-tris carbaz, 4-carbazol-9-yl-n,n-bis 4-carbazol-9-ylphenyl aniline
PubChem CID 9962045
IUPAC Name 4-(9H-carbazol-9-yl)-N,N-bis[4-(9H-carbazol-9-yl)phenyl]aniline
SMILES C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=C(C=C1)N(C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 266°C
Color White-Yellow
Quantity 1 g
Formula Weight 740.91
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′,4′′-Tri-9-carbazolyltriphenylamine