missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(4-Chlorophenyl)-1H-pyrazol-1-yl]piperidine, Thermo Scientific Chemicals

Catalog No. AAH5080403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5080403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5080403 Supplier Thermo Scientific Chemicals Supplier No. H5080403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 902836-38-8 | Molecular Formula: C14H16ClN3 | Molecular Weight: 261.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 902836-38-8
Molecular Formula C14H16ClN3
Molecular Weight (g/mol) 261.75
MDL Number MFCD08060997
InChI Key QPXZSTSEYCYKLI-UHFFFAOYSA-N
Synonym 4-4-4-chlorophenyl-1h-pyrazol-1-yl piperidine, 4-4-4-chlorophenyl pyrazol-1-yl piperidine, acmc-20amql, 4-4-chlorophenyl-1-4-piperidyl pyrazole, 4-4-4-chlorophenyl-1h-pyrazol-1-ylpiperidine, 4-4-4-chloro-phenyl-pyrazol-1-yl-piperidine
PubChem CID 24212026
IUPAC Name 4-[4-(4-chlorophenyl)pyrazol-1-yl]piperidine
SMILES ClC1=CC=C(C=C1)C1=CN(N=C1)C1CCNCC1

Specifications

Quantity 1 g
Formula Weight 261.76
Chemical Name or Material 4-[4-(4-Chlorophenyl)-1H-pyrazol-1-yl]piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only