missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™
Supplier: TCI America T23495G
Specifications
| 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | |
| 151°C | |
| C11H16BNO2 | |
| 5 g | |
| NLTIETZTDSJANS-UHFFFAOYSA-N | |
| 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | |
| 2734648 | |
| ≥98.0% (GC,T) |
| 181219-01-2 | |
| White-Yellow | |
| MFCD01319051 | |
| 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 4-pyridineboronic acid pinacol ester, pyridine-4-boronic acid pinacol ester, 4-pyridylboronic acid pinacol ester, pyridine-4-boronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-4-boronic pinacolate, pyridine-4-boronic acid pinacol cyclic ester, 2-4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine-4-boronic pinacol ester | |
| CC1(C)OB(OC1(C)C)C1=CC=NC=C1 | |
| 205.06 | |
| 205.06 | |
| Crystalline Powder |
Chemical Identifiers
| 181219-01-2 | |
| 205.06 | |
| NLTIETZTDSJANS-UHFFFAOYSA-N | |
| 2734648 | |
| CC1(C)OB(OC1(C)C)C1=CC=NC=C1 |
| C11H16BNO2 | |
| MFCD01319051 | |
| 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 4-pyridineboronic acid pinacol ester, pyridine-4-boronic acid pinacol ester, 4-pyridylboronic acid pinacol ester, pyridine-4-boronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-4-boronic pinacolate, pyridine-4-boronic acid pinacol cyclic ester, 2-4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine-4-boronic pinacol ester | |
| 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Safety and Handling
TSCA : No