missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzothiophene 98.0+%, TCI America™

Supplier:  TCI America T32365G

 View more versions of this product

Catalog No. T32365G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzothiophene
70°C
C18H19BO2S
5 g
XPRBSZLRNLZXNT-UHFFFAOYSA-N
2-dibenzothiophen-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
57417006
≥98.0% (GC,T)
912824-84-1
White-Yellow
MFCD18452170
4-Dibenzothiopheneboronic Acid Pinacol Ester, 2-(Dibenzothiophene-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C3C(=CC=C2)C4=CC=CC=C4S3
310.218
310.22
Crystalline Powder

Chemical Identifiers

912824-84-1
310.218
XPRBSZLRNLZXNT-UHFFFAOYSA-N
57417006
B1(OC(C(O1)(C)C)(C)C)C2=C3C(=CC=C2)C4=CC=CC=C4S3
C18H19BO2S
MFCD18452170
4-Dibenzothiopheneboronic Acid Pinacol Ester, 2-(Dibenzothiophene-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-dibenzothiophen-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions