missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzothiophene 98.0+%, TCI America™
Supplier: TCI America T32361G
Specifications
| 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzothiophene | |
| 70°C | |
| C18H19BO2S | |
| 1 g | |
| XPRBSZLRNLZXNT-UHFFFAOYSA-N | |
| 2-dibenzothiophen-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 57417006 | |
| ≥98.0% (GC,T) |
| 912824-84-1 | |
| White-Yellow | |
| MFCD18452170 | |
| 4-Dibenzothiopheneboronic Acid Pinacol Ester, 2-(Dibenzothiophene-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| B1(OC(C(O1)(C)C)(C)C)C2=C3C(=CC=C2)C4=CC=CC=C4S3 | |
| 310.218 | |
| 310.22 | |
| Crystalline Powder |
Chemical Identifiers
| 912824-84-1 | |
| 310.218 | |
| XPRBSZLRNLZXNT-UHFFFAOYSA-N | |
| 57417006 | |
| B1(OC(C(O1)(C)C)(C)C)C2=C3C(=CC=C2)C4=CC=CC=C4S3 |
| C18H19BO2S | |
| MFCD18452170 | |
| 4-Dibenzothiopheneboronic Acid Pinacol Ester, 2-(Dibenzothiophene-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 2-dibenzothiophen-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No