missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran 99.0+%, TCI America™

Catalog No. p-7137814
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T36021G 1 g
T36025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T36021G Supplier TCI America Supplier No. T36021G

Chemical Identifiers

CAS 912824-85-2
Molecular Formula C18H19BO3
Molecular Weight (g/mol) 294.16
MDL Number MFCD18910632
InChI Key AVWYETCDUMCCNK-UHFFFAOYSA-N
Synonym Dibenzofuran-4-boronic Acid Pinacol Ester, 2-(Dibenzofuran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 60146103
IUPAC Name 4,4,5,5-tetramethyl-2-{8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-6-yl}-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC2=C1OC1=C2C=CC=C1

Specifications

Melting Point 109°C
Color White
Quantity 1 g
Formula Weight 294.16
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran