missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%

Catalog No. p-2690473
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC366390010 1 g Glass bottle
AC366390050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC366390010 Supplier Thermo Scientific Chemicals Supplier No. 366390010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 180516-87-4 | C13H17BO4 | 248.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 180516-87-4
Molecular Formula C13H17BO4
Molecular Weight (g/mol) 248.09
MDL Number MFCD01863710
InChI Key IYDKBQIEOBXLTP-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 4-carboxyphenylboronic acid pinacol ester, 4-carboxylphenylboronic acid pinacol ester, 4-carboxyphenylboronic acid, pinacol ester, 4-carboxyphenylboronicacidpinacolester, 4-carboxybenzeneboronic acid pinacol ester, 4-carboxyphenylboronic acid pinacolate, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 4-carboxyphenylboronic acid,pinacol ester, 4-carboxyphenyl boronic acid, pinacol ester
PubChem CID 2734621
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 227.0°C to 232.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 248.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
If skin irri
Recommended Storage Refrigerator +4°C

RUO – Research Use Only