missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 98.0+%, TCI America™

Catalog No. p-7137036
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19511G 1 g
T19515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19511G Supplier TCI America Supplier No. T19511G
Only null left

Chemical Identifiers

CAS 214360-73-3
Molecular Formula C12H18BNO2
Molecular Weight (g/mol) 219.09
MDL Number MFCD02093721
InChI Key ZANPJXNYBVVNSD-UHFFFAOYSA-N
Synonym 4-aminophenylboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-aminophenylboronic acid pinacolate, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, pinacol cyclic ester, 2-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-aminophenylboronic acid, pinacol ester, 4-aminophenylboronicacidpinacolester, 4-aminobenzeneboronic acid pinacol ester
PubChem CID 2734620
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(N)C=C1

Specifications

Melting Point 170°C
Quantity 1 g
Formula Weight 219.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline