missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%

Catalog No. p-2690470
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC366380010 1 g Glass bottle
AC366380050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC366380010 Supplier Thermo Scientific Chemicals Supplier No. 366380010
Only null left

CAS: 214360-73-3 | C12H18BNO2 | 219.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 214360-73-3
Molecular Formula C12H18BNO2
Molecular Weight (g/mol) 219.09
MDL Number MFCD02093721
InChI Key ZANPJXNYBVVNSD-UHFFFAOYSA-N
Synonym 4-aminophenylboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-aminophenylboronic acid pinacolate, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, pinacol cyclic ester, 2-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-aminophenylboronic acid, pinacol ester, 4-aminophenylboronicacidpinacolester, 4-aminobenzeneboronic acid pinacol ester
PubChem CID 2734620
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(N)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 165°C to 170°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 219.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage product may turn dark on exposure to air

RUO – Research Use Only