missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyran 98.0+%, TCI America™

Catalog No. T32171G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T32171G Supplier TCI America Supplier No. T32171G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 287944-16-5
Molecular Formula C11H19BO3
Molecular Weight (g/mol) 210.08
MDL Number MFCD11052631
InChI Key DOSGEBYQRMBTGS-UHFFFAOYSA-N
Synonym 3,6-dihydro-2h-pyran-4-boronic acid pinacol ester, 2-3,6-dihydro-2h-pyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-pyran, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 2h-pyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 3,6-dihydro-2h-pyran-4-boronic acid, pinacol ester, 3,6-dihydro-2h-pyran-4-boronicacidpinacolester, 3,6-dihydro-2h-pyran-4-boronic acid pinacolester, 3,6 dihydro-2h-pyran-4-boronic acid pinacol ester
PubChem CID 11218053
IUPAC Name 2-(3,6-dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CCOCC1

Specifications

Melting Point 63°C
Quantity 1 g
Formula Weight 210.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyran
TSCA No
Recommended Storage Refrigerator