missing translation for 'onlineSavingsMsg'
Learn More

4,4,4,4-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid), ≥95.0%, MilliporeSigma™ Supelco™

Catalog No. 111014004
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014004 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014004 Supplier Merck Emd Millipore Supplier No. 89456100MGF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00070632
Synonym meso-Tetraphenylporphine-4,4 ′,4 ′′,4 ′′′-tetrasulfonic acid

Specifications

CAS 35218-75-8
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C44H30N4O12S4
Quantity 100 mg
Formula Weight 934.99
Percent Purity ≥95.0% (HPLC); ≥95.0%
RTECSNumber DB7339000