missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Trifluoromethyl)phenyl]-4-piperidinol, 97%, Thermo Scientific™

Catalog No. AC136980250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC136980250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC136980250 Supplier Thermo Scientific Chemicals Supplier No. 136980250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2249-28-7
Molecular Formula C12H14F3NO
Molecular Weight (g/mol) 245.245
MDL Number MFCD00044832
InChI Key ILKPZCKFWLTEBQ-UHFFFAOYSA-N
Synonym 4-3-trifluoromethyl phenyl-4-piperidinol, 4-3-trifluoromethyl phenyl piperidin-4-ol, 4-piperidinol, 4-3-trifluoromethyl phenyl, 4-alpha,alpha,alpha-trifluoro-m-tolyl piperidin-4-ol, 4-3-trifuoromethyl phenyl-4-piperidinol, 4-3-trifluoromethylphenyl-4-piperidinol, 4-pound 3-trifluoromethylphenyl-4-piperidinol, 4-3-trifluoromethylphenyl-4'-piperidinol, 4-3-trifluoromethyl phenyl-4'-piperidinol
PubChem CID 75255
IUPAC Name 4-[3-(trifluoromethyl)phenyl]piperidin-4-ol
SMILES C1CNCCC1(C2=CC(=CC=C2)C(F)(F)F)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 91°C to 95°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Formula Weight 245.24
Percent Purity ≥96.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-[3-(Trifluoromethyl)phenyl]-4-piperidinol, 97%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only