missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl Bromide 95.0+%, TCI America™

Catalog No. p-7137435
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2819200MG 200 mg
T28191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2819200MG Supplier TCI America Supplier No. T2819200MG

Chemical Identifiers

CAS 92367-11-8
Molecular Formula C9H6BrF3N2
Molecular Weight (g/mol) 279.06
MDL Number MFCD19442581
InChI Key BCFUQXOZHPIAPS-UHFFFAOYSA-N
Synonym 3-[4-(Bromomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine
PubChem CID 10826496
IUPAC Name 3-[4-(bromomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine
SMILES FC(F)(F)C1(N=N1)C1=CC=C(CBr)C=C1

Specifications

Quantity 200 mg
UN Number 3265
Formula Weight 279.06
Percent Purity ≥95.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl Bromide
TSCA No
Recommended Storage Refrigerator