missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl Alcohol 97.0+%, TCI America™
$160.30 - $546.17
Chemical Identifiers
| CAS | 87736-88-7 |
|---|---|
| Molecular Formula | C9H7F3N2O |
| Molecular Weight (g/mol) | 216.163 |
| InChI Key | UZHHTEASLDJYBO-UHFFFAOYSA-N |
| Synonym | 3-[4-(Hydroxymethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine |
| PubChem CID | 11367905 |
| IUPAC Name | [4-[3-(trifluoromethyl)diazirin-3-yl]phenyl]methanol |
| SMILES | C1=CC(=CC=C1CO)C2(N=N2)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T2818200MG
|
TCI America
T2818200MG |
200 mg |
Each for $160.30
|
|
|||||
|
T28181G
|
TCI America
T28181G |
1 g |
Each for $546.17
|
|
|||||
Chemical Identifiers
| 87736-88-7 | |
| 216.163 | |
| 3-[4-(Hydroxymethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine | |
| [4-[3-(trifluoromethyl)diazirin-3-yl]phenyl]methanol |
| C9H7F3N2O | |
| UZHHTEASLDJYBO-UHFFFAOYSA-N | |
| 11367905 | |
| C1=CC(=CC=C1CO)C2(N=N2)C(F)(F)F |
Specifications
| 87736-88-7 | |
| C9H7F3N2O | |
| 3-[4-(Hydroxymethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine | |
| C1=CC(=CC=C1CO)C2(N=N2)C(F)(F)F | |
| 216.163 | |
| 216.16 | |
| Liquid |
| Yellow | |
| 200 mg | |
| UZHHTEASLDJYBO-UHFFFAOYSA-N | |
| [4-[3-(trifluoromethyl)diazirin-3-yl]phenyl]methanol | |
| 11367905 | |
| ≥97.0% (HPLC) | |
| 4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl Alcohol |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator