Learn More
4-(3-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride, 97%
CAS: 874839-13-1 | C12H7ClF3NO3S | 337.697 g/mol
$61.96 - $443.10
Chemical Identifiers
| CAS | 874839-13-1 |
|---|---|
| Molecular Formula | C12H7ClF3NO3S |
| Molecular Weight (g/mol) | 337.697 |
| MDL Number | MFCD07774164 |
| InChI Key | FDAYBQVWHDJJOL-UHFFFAOYSA-N |
| Synonym | 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-3-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, chloro 4-3-trifluoromethyl 2-pyridyloxy phenyl sulfone, 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride |
| PubChem CID | 45594320 |
| IUPAC Name | 4-[3-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride |
| SMILES | C1=CC(=C(N=C1)OC2=CC=C(C=C2)S(=O)(=O)Cl)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH33473MD
|
Thermo Scientific Chemicals
H33473MD |
250 mg |
N/A
|
|
|||||
|
AAH3347306
|
Thermo Scientific Chemicals
H3347306 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 874839-13-1 | |
| 337.697 | |
| FDAYBQVWHDJJOL-UHFFFAOYSA-N | |
| 45594320 | |
| C1=CC(=C(N=C1)OC2=CC=C(C=C2)S(=O)(=O)Cl)C(F)(F)F |
| C12H7ClF3NO3S | |
| MFCD07774164 | |
| 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-3-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, chloro 4-3-trifluoromethyl 2-pyridyloxy phenyl sulfone, 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride | |
| 4-[3-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride |
Specifications
| 874839-13-1 | |
| C12H7ClF3NO3S | |
| 250 mg | |
| 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-3-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, chloro 4-3-trifluoromethyl 2-pyridyloxy phenyl sulfone, 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride | |
| C1=CC(=C(N=C1)OC2=CC=C(C=C2)S(=O)(=O)Cl)C(F)(F)F | |
| 337.697 | |
| 337.71 | |
| 4-(3-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride |
| 63°C to 65°C | |
| MFCD07774164 | |
| UN3261 | |
| FDAYBQVWHDJJOL-UHFFFAOYSA-N | |
| 4-[3-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride | |
| 45594320 | |
| 97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only