missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Thienyl)benzaldehyde, ≥97%, Thermo Scientific™

Chemical Identifiers

CAS 157730-74-0
Molecular Formula C11H8OS
Molecular Weight (g/mol) 188.244
MDL Number MFCD04039151
InChI Key MGPZYZLJJSEFLU-UHFFFAOYSA-N
Synonym 4-thiophen-3-yl benzaldehyde, 4-3-thienyl benzaldehyde, 4-3-thienyl benzenecarbaldehyde, 4-thiophen-3-yl-benzaldehyde, benzaldehyde,4-3-thienyl, 4-thien-3-yl benzaldehyde, pubchem15939, thienylbenzenecarbaldehyde, 4-3-thiophenyl benzaldehyde, benzaldehyde, 4-3-thienyl
PubChem CID 3280631
IUPAC Name 4-thiophen-3-ylbenzaldehyde
SMILES C1=CC(=CC=C1C=O)C2=CSC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC48704CB
View Documents
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC48704DA
View Documents
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC48704EA
View Documents
N/A 10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

157730-74-0
188.244
MGPZYZLJJSEFLU-UHFFFAOYSA-N
3280631
C1=CC(=CC=C1C=O)C2=CSC=C2
C11H8OS
MFCD04039151
4-thiophen-3-yl benzaldehyde, 4-3-thienyl benzaldehyde, 4-3-thienyl benzenecarbaldehyde, 4-thiophen-3-yl-benzaldehyde, benzaldehyde,4-3-thienyl, 4-thien-3-yl benzaldehyde, pubchem15939, thienylbenzenecarbaldehyde, 4-3-thiophenyl benzaldehyde, benzaldehyde, 4-3-thienyl
4-thiophen-3-ylbenzaldehyde

Specifications

157730-74-0
100.0
Amber glass bottle
MFCD04039151
MGPZYZLJJSEFLU-UHFFFAOYSA-N
4-thiophen-3-ylbenzaldehyde
3280631
97%
96.5
97%
C11H8OS
4-thiophen-3-yl benzaldehyde, 4-3-thienyl benzaldehyde, 4-3-thienyl benzenecarbaldehyde, 4-thiophen-3-yl-benzaldehyde, benzaldehyde,4-3-thienyl, 4-thien-3-yl benzaldehyde, pubchem15939, thienylbenzenecarbaldehyde, 4-3-thiophenyl benzaldehyde, benzaldehyde, 4-3-thienyl
C1=CC(=CC=C1C=O)C2=CSC=C2
188.244
188.25
4-(3-Thienyl)benzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents