missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Phenylpropyl)pyridine 98.0+%, TCI America™

Catalog No. p0218500g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
P0218500G 500 g
P021825G 25 g
2 options

Catalog No. P0218500G

Supplier: TCI America P0218500G

Only null left

Chemical Identifiers

CAS 2057-49-0
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
MDL Number MFCD00047458
InChI Key AQIIVEISJBBUCR-UHFFFAOYSA-N
Synonym 4-3-phenylpropyl pyridine, pyridine, 4-3-phenylpropyl, 1-phenyl-3-4-pyridyl propane, unii-yw9q68ek6b, 4-phenylpropylpyridine, yw9q68ek6b, 1-4-pyridyl-3-phenylpropane, acmc-1ccts, dsstox_cid_24869, dsstox_rid_80544
PubChem CID 74937
IUPAC Name 4-(3-phenylpropyl)pyridine
SMILES C1=CC=C(C=C1)CCCC2=CC=NC=C2

Specifications

Color Yellow
Quantity 500 g
Formula Weight 197.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(3-Phenylpropyl)pyridine
EINECSNumber (5)-3720
TSCA Yes