missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Phenylpropyl)pyridine, 98%

Catalog No. AAA1359030
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1359030 250 g
AAA1359018 50 g
AAA135900B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1359030 Supplier Thermo Scientific Chemicals Supplier No. A1359030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2057-49-0 | C14H15N | 197.281 g/mol

4-(3-Phenylpropyl)pyridine is used in electrochemical processes for a novel system of microdroplets of PPP immobilized at graphite and in mesoporous ceramic electrodes, PPP has been used to investigate electrochemically driven mono-, di-, and tri-carboxylate anion transfer at the 4-(3-phenylpropyl)pyridine/aqueous electrolyte interface for a triple phase boundary system at graphite electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(3-Phenylpropyl)pyridine is used in electrochemical processes for a novel system of microdroplets of PPP immobilized at graphite and in mesoporous ceramic electrodes, PPP has been used to investigate electrochemically driven mono-, di-, and tri-carboxylate anion transfer at the 4-(3-phenylpropyl)pyridine/aqueous electrolyte interface for a triple phase boundary system at graphite electrodes.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 2057-49-0
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
MDL Number MFCD00047458
InChI Key AQIIVEISJBBUCR-UHFFFAOYSA-N
Synonym 4-3-phenylpropyl pyridine, pyridine, 4-3-phenylpropyl, 1-phenyl-3-4-pyridyl propane, unii-yw9q68ek6b, 4-phenylpropylpyridine, yw9q68ek6b, 1-4-pyridyl-3-phenylpropane, acmc-1ccts, dsstox_cid_24869, dsstox_rid_80544
PubChem CID 74937
IUPAC Name 4-(3-phenylpropyl)pyridine
SMILES C1=CC=C(C=C1)CCCC2=CC=NC=C2

Specifications

Density 1.029
Boiling Point 320°C to 322°C
Flash Point 140°C (284°F)
Refractive Index 1.563
Quantity 250 g
Beilstein 142242
Solubility Information Difficult to mix.
Formula Weight 197.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-(3-Phenylpropyl)pyridine
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement

H302

Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 218-159-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only